Order Entry
Czech Republic
ContactUsLinkComponent
Potassium dihydrogen phosphate Ph. Eur., NF, BP endotoxin tested
Potassium dihydrogen phosphate Ph. Eur., NF, BP endotoxin tested
Supplier:  VWR Chemicals
Certificates
undefined
Potassium dihydrogen phosphate Ph. Eur., NF, BP endotoxin tested
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    Conforms (see CoA/CoS)
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Bacterial endotoxins:
    Max. 2.5 EU/g
  • Residual solvents:
    Passes test
  • Residues of metal catalysts or reagents:
    Unlikely by manuf.process
  • Cd (Cadmium):
    Max. 1 ppm
  • Co (Cobalt):
    Max. 1 ppm
  • Hg (Mercury):
    Max. 3 ppm
  • Ni (Nickel):
    Max. 1 ppm
  • Pb (Lead):
    Max. 3 ppm
  • V (Vanadium):
    Max. 4 ppm
  • Conforms to Ph.Eur./NF/BP:
    Passes test
  • Ph.Eur./BP specifications::
    Max. 10 ppm
  • Assay (dried substance) [Ph.Eur./BP]:
    98.0 --> 100.5 %
  • Identification A:
    Passes test Ph.Eur./BP
  • Identification B:
    Passes test Ph.Eur./BP
  • Identification C:
    Passes test Ph.Eur./BP
  • Appearance of solution:
    Passes test Ph.Eur./BP
  • pH (20°C; 5 %) [Ph.Eur./BP]:
    4.2 --> 4.5
  • Reducing substances:
    Passes test Ph.Eur./BP
  • Cl (Chloride) [Ph.Eur./BP]:
    Max. 200 ppm
  • SO4 (Sulphate) [Ph.Eur./BP]:
    Max. 300 ppm
  • As (Arsenic) [Ph.Eur./BP]:
    Max. 2 ppm
  • Fe (Iron) [Ph.Eur./BP]:
    Max. 10 ppm
  • Loss on drying (125-130°C) [Ph.Eur./BP]:
    Max. 2.0 %
  • Assay (dried basis) [NF]:
    98.0 --> 100.5 %
  • As (Arsenic) [NF]:
    Max. 3 μg/g
  • Pb (Lead) [NF]:
    Max. 5 μg/g
  • Limit of fluoride [NF]:
    Max. 10 μg/g
  • Insoluble substances [NF]:
    Max. 0.2 %
  • Loss on drying (105°C; 4 h) [NF]:
    Max. 1.0 %

Specification Test Results

Frequently Bought Together