Anhydrotetracycline hydrochloride
Catalog # J66688.MA
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 mg
- Environmentally Preferable:
Specifications
About this item
:
:462.883 g/mol
Melting Pt:220…223 °C
Storage Temperature:Freezer
CAS Number:13803-65-1
Formula:C₂₂H₂₃ClN₂O₇
MDL Number:MFCD00151453