

Soap solution according to boutron and boudet solution DIN 8106, Fluka™
Catalog # 35366-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Factor:0.99 - 1.01
Specification Test Results
About this item
:
Boiling Pt:78 °C (1.013 hPa)
Density:0.93 g/cm3 (20 °C)
Flash Pt:22 °C
Storage Temperature:Ambient
UN:1170
ADR:3,II
:GHS07
:GHS02
MDL Number:MFCD00148418