2,2,4-Trimethylpentane, anhydrous (max. 0.005% H₂O)
:540-84-1
:
:(CH₃)₂CHCH₂C(CH₃)₃
:MFCD00008943
:114.23 g/mol
:13,05212
:99.2 °C (1013 hPa)
:–107 °C
:0.692 g/cm³ (20 °C)
:–12 °C
:208-759-1
:1262
:3,II
:GHS02
:GHS08
:GHS07
:GHS09
:Ambient
- Assay (GC, on anhydrous basis):99.5-100 %
- Residue after evaporation:0-0.001 % w/w
- Water (KF):0-0.005 % w/w
- Acidity (as Acetic acid):0-0.003 %
Frequently Bought Together