Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, Piroxicam, Ph.Eur., RS
Catalog # EDQMP2130000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Piroxicam, Ph.Eur., RS
Catalog # EDQMP2130000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    Reference standard
  • CAS No.:
    36322-90-4
  • Description:
    Piroxicam
  • Formula:
    C₁₅H₁₃N₃O₄S
  • Storage temperature:
    Ambient
  • Description:
    Piroxicam

Specifications

About this item

  • In Monograph 0944
  • Solubility in water: less than 0,1 g/l