Order Entry
Czech Republic
ContactUsLinkComponent
Hydrochloric acid 5 mol/l (5 N) volumetric solution
Hydrochloric acid 5 mol/l (5 N) volumetric solution
Catalog # APPC182109.1211
Supplier:  PanReac AppliChem
CAS Number:  
Hydrochloric acid 5 mol/l (5 N) volumetric solution
Catalog # APPC182109.1211
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer at 20 °C:
    0,999 - 1,001
  • Traceability:
    NIST
  • Density at 20/4:
    1,081 - 1,085 g/ml
  • Appearance:
    Clear colourless liquid
  • Solubility:
    Miscible with water
  • Storage:
    RT

Specification Test Results

About this item

 :
ADR:8,II
 :36.46 g/mol
UN:1789
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324