
Kyselina citronová monohydrát USP, BP, Ph. Eur., JP, pure
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:5949-29-1
:
Formula:HOC(COOH)(CH₂COOH)₂·H₂O
MDL Number:MFCD00149972
Melting Pt:135 °C
EINECS:201-069-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (Acidim.) calc. a.a.s.:99,5 - 100,5%
- Identity according to Pharmacopoeias:Passes test
- Appearance of solution:Passes test
- Insoluble matter in H₂O:0,01%
- Darkened substances by H₂SO₄:Passes test
- Residue on ignition (as SO₄):0,05%
- Chloride (Cl) :0,005%
- Sulfate (SO₄) :0,01%
- Colour of solution:Passes test
- Residual solvents (Ph.Eur/USP):Passes test
- Oxalate (C₂O₄) :0,035%
- Water (H₂O):7,5 - 8,8%
- Heavy metals (as Pb):0,001%
- Barium (Ba):Passes test
- Calcium (Ca):0,02%
- Magnesium (Mg):0,005%
- Cadmium (Cd):0,5 ppm
- Lead (Pb):0,5 ppm
- Arsenic (As):1 ppm
- Mercury (Hg):1,5 ppm
- Cobalt (Co):5 ppm
- Vanadium (V):10 ppm
- Nickel (Ni):20 ppm
- Thallium (Tl):5 ppm
- Gold (Au):10 ppm
- Palladium (Pd):10 ppm
- Iridium (Ir):10 ppm
- Osmium (Os):10 ppm
- Rhodium (Rh):10 ppm
- Ruthenium (Ru):10 ppm
- Selenium (Se):15 ppm
- Silver (Ag):15 ppm
- Platinum (Pt):10 ppm
- Lithium (Li):55 ppm
- Antimony (Sb):120 ppm
- Molybdenum (Mo):25 ppm
- Copper (Cu):250 ppm
- Tin (Sn):600 ppm
- Chromium (Cr):25 ppm
Specification Test Results
Frequently Bought Together