ContactUsLinkComponent
Human bone marrow, unprocessed
  BM008F-4
 :  
Human bone marrow, unprocessed
  BM008F-4
 :  
  • Cell Format:
    Fresh
  • Pk:
    100 mL
  • Single Donor/Pooled:
    Pooled
  • Description:
    Human unprocessed bone marrow
  • Seq:
    03
Bone marrow is a major source of stem cells, which are being used in cancer research, cell studies, transplantation studies, and other research.
  • Test negative for Hepatitis B and C, HIV 1/2, HTLV- I/II, HCV/HBV, WNV, Trypanasoma cruzi, and Syphilis
Bone marrow collected from healthy human volunteer donors under IRB approved informed consent, drawn from the posterior iliac crest using a syringe containing anticoagulant.
 : Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.