Mesna, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
- Catalog No:
- 1392807.
- 1392818.
- 1392829.
- Description:
- Mesna
- Mesna Related Compound A
- Mesna Related Compound B
- Synonym(s):
- 2-(Acetylthio)ethane-1-sulfonic acid, potassium salt, crystal adduct with potassium chloride
- 2,2'-Disulfanediylbis(ethane-1-sulfonic acid), dipotassium salt, crystal adduct with sodium chloride
- CAS No.:
- 19767-45-4
- 45127-11-5
- Formula:
- C₂H₅NaO₃S₂
- C₄H₁₀O₆S₄
Product Family Options
- DescriptionStandardsPack typePk
- MesnaUSP reference standardVial200 mg1 * 200 mg8 470,00 Kč
Specifications
Cat. No.1392807.DescriptionMesnaSynonym(s)CAS No.19767-45-4FormulaC₂H₅NaO₃S₂ - Mesna Related Compound AUSP reference standardVial50 mg1 * 50 mg28 460,00 Kč
Specifications
Cat. No.1392818.DescriptionMesna Related Compound ASynonym(s)2-(Acetylthio)ethane-1-sulfonic acid, potassium salt, crystal adduct with potassium chlorideCAS No.Formula - Mesna Related Compound BUSP reference standardVial50 mg1 * 50 mg28 460,00 Kč
Specifications
Cat. No.1392829.DescriptionMesna Related Compound BSynonym(s)2,2'-Disulfanediylbis(ethane-1-sulfonic acid), dipotassium salt, crystal adduct with sodium chlorideCAS No.45127-11-5FormulaC₄H₁₀O₆S₄
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



