Order Entry
Czech Republic
ContactUsLinkComponent
Lamivudine, Reference Standards
Lamivudine, Reference Standards
Supplier:  USP
Certificates
Lamivudine, Reference Standards
Supplier:  USP

Specifications

  • CAS No.:
    134678-17-4
  • Formula:
    C₈H₁₁N₃O₃S
  • Description:
    Lamivudine

Specifications

Frequently Bought Together