Order Entry
Czech Republic
ContactUsLinkComponent
Economy lockout
Economy lockout
Catalog # BRDY236920
Supplier:  Brady
undefined
Economy lockout
Catalog # BRDY236920
Supplier:  Brady

Specifications

  • Type:
    Economy lockout
  • Colour:
    Red
  • Pk:
    1

Specifications

About this item

This economy lockout is ideal for use on fuse or switch boxes.

  • Made from sturdy steel
  • Six padlocks can be applied

Max. shackle diameter: 10,00 mm

Min. shackle clearance: 20,00 mm