Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant Carbonic Anhydrase 3 (from E. coli)
Human recombinant Carbonic Anhydrase 3 (from E. coli)
Catalog # PRSI92-083
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Carbonic Anhydrase 3 (from E. coli)
Catalog # PRSI92-083
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Carbonic Anhydrase 3
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    30.6 kD
  • Sequence:
    Ala2-Lys260
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM Tris, 150 mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Carbonic Anhydrase 3 (CA3) belongs to the Alpha-Carbonic Anhydrase family that encodes carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyse the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and it is present at high levels in skeletal muscle with much lower levels found in cardiac and smooth muscle. CA3 is activated by proton donors such as imidazole and the dipeptide histidylhistidine. CA3 is inhibited by coumarins and sulfonamide derivatives such as acetazolamide.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.