Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant GGPPSase (from E. coli)
Human recombinant GGPPSase (from E. coli)
Catalog # PRSI91-209
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant GGPPSase (from E. coli)
Catalog # PRSI91-209
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    GGPPSase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    37 kD
  • Sequence:
    Met1-Glu300
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 8.0 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Geranylgeranyl pyrophosphate synthase (GGPS1) is a member of the FPP/GGPP synthase family. GGPS1 is highly expressed in testis, heart and skeletal muscle. GGPS1 is localised in the cytoplasm and has geranylgeranyl diphosphate (GGPP) synthase activity. It catalyses the trans-addition of the three molecules of IPP onto DMAPP to form geranylgeranyl pyrophosphate, an important precursor of carotenoids and geranylated proteins. Other transcriptional splice variants have been found.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.