Order Entry
Czech Republic
ContactUsLinkComponent
 
Human recombinant Carbonic Anhydrase 13 (from E. coli)
Catalog # PRSI91-091
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Carbonic Anhydrase 13 (from E. coli)
Catalog # PRSI91-091
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Carbonic Anhydrase 13
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    30.51 kD
  • Sequence:
    Met1-His262
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Carbonic Anhydrase 13 (CA13) belongs to the carbonic anhydrase family which can catalyses the reversible hydration recation of carbon dioxide. Carbonic anhydrases participate in many biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. CA13 is a cytosolic enzyme and is widely expressed in human, such as thymus, small intestine, spleen, prostate, ovary, colon and testis, indicating that it may play a key role in several organs. CA13 is inhibited by acetazolamide.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.