Order Entry
Czech Republic
ContactUsLinkComponent
Mouse recombinant IL-21R non-lytic (from CHO cells)
Mouse recombinant IL-21R non-lytic (from CHO cells)
Catalog # PRSI90-513
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant IL-21R non-lytic (from CHO cells)
Catalog # PRSI90-513
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Biological activity:
    Shows the biological function of the IL-21R moiety and exerts a prolonged circulating half-life caused by the modified Fc domain.
  • Protein synonyms:
    CD360|Interleukin-21 Receptor|NILR
  • Protein/peptide name:
    IL-21R non-lytic
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    20-234
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.

Specifications

About this item

Interleukin-21 receptor (IL-21R) is a type I transmembrane glycoprotein within the class I cytokine receptor family, type 4 subfamily. Complex formation between IL-21R and the common gamma chain (gammac) is required for signalling. IL-21R is expressed mainly on B cells (highest on mature, activated, follicular and germinal center B cells), NK cells and activated T cells, but is also found on dendritic cells, alternatively activated macrophages, intestinal lamina propria fibroblasts and epithelial cells and keratinocytes. B cell IL-21R engagement induces Blimp1 (which mediates plasma cell differentiation) and is important for memory responses. IL-21R engagement on mouse NK cells enhances their cytotoxic activity and IFN-gamma production. IL-21R engagement on CD8+ T cells aids control of viral infection and tumour growth; IL-21R is also necessary for sufficient numbers of regulatory T cells to combat chronic inflammation. IL-21R expression is often upregulated in allergic skin inflammation, systemic lupus erythematosus and diffuse large B cell lymphoma (DLBCL).

Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.