Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant IL-21 mutant (from E. coli)
Human recombinant IL-21 mutant (from E. coli)
Catalog # PRSI90-510
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL-21 mutant (from E. coli)
Catalog # PRSI90-510
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein synonyms:
    Interleukin-21
  • Protein/peptide name:
    IL-21 mutant
  • Purity:
    >90% (SDS-PAGE)
  • Sequence:
    32-162
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.

Specifications

About this item

Interleukin-21 (IL-21) is a key factor in the transition between innate and adaptive immune responses secreted by activated T cells. The IL-21 receptor (IL-21R) is expressed in lymphoid tissue, in particular by NK, B, T and dendritic cells, macrophages and endothelial cells. Recent evidence suggests that IL-21 plays a supportive role in the proliferation of T and B cells and influences the cytolytic activity of natural killer cells. IL-21 has been shown to up-regulate genes associated with innate immunity and to inhibit the differentiation of naive T helper cells. IL-21 specifically inhibits IFN-gamma production from developing TH1 cells and is preferentially expressed by TH2 cells. Furthermore IL-21 has been identified as a growth and survival factor for human myeloma cells. IL-21/IL-21R interactions have a unique role in sequentially activating both innate and adaptive immune responses against poorly immunogenic tumours, leading to tumour rejection that is perforin dependent but IFN-gamma independent.

Fusion-Tag: His Tag

This recombinant proteins is for research use only.