Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant BAFF (active) (soluble) (from E. coli)
Human recombinant BAFF (active) (soluble) (from E. coli)
Catalog # PRSI90-294
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant BAFF (active) (soluble) (from E. coli)
Catalog # PRSI90-294
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at −20 °C.
  • Biological activity:
    Increases B cell survival/proliferation. Increases CD21/CD23 expression on B cells in vivo. Activates BAFF-R, TACI and BCMA receptors. Works at concentrations <20ng/ml.
  • Protein synonyms:
    B Cell Activating Factor|CD257|TALL-1|BLyS|Tumor Necrosis Factor Ligand Superfamily Member 13B|TNFSF13B
  • Protein/peptide name:
    BAFF (active) (soluble)
  • Purity:
    >95% (SDS-PAGE)
  • Sequence:
    134-285
  • Formulation:
    Lyophilized, Contains PBS.

Specifications

About this item

BAFF is mainly produced by innate immune cells such as neutrophils, monocytes, macrophages, dendritic cells, follicular dendritic cells. T cells, activated B cells, some malignant B cells and also non-lymphoid cells like astrocytes, synoviocytes and epithelial cells can also produce BAFF. BAFF binds three distinct receptors (BAFF-R, TACI and BCMA) expressed predominantly on B cells, although activated T cells also express BAFF-R. BAFF is a master regulator of peripheral B cell survival, and together with IL-6, promotes Ig class-switching and plasma cell differentiation. Besides its major role in B cell biology, BAFF co-stimulates activated T cells. Deregulated expression of BAFF leads to autoimmune disorders in mice. In humans, elevated levels of soluble BAFF have been detected in the serum of patients with various autoimmune diseases such as Sjoegren syndrome, Rheumatoid arthritis (RA), Multiple sclerosis (MS) and Systemic Lupus Erythematosus (SLE). BAFF has also increased levels in some lymphoid cancers. Processed human BAFF can either remain as a trimer, which is usual for TNF family ligands or assemble into 60-mer composed of 20 trimers. Mouse BAFF 60-mer has been identified in the serum of BAFF transgenic mice. Oligomerisation of BAFF 3-mer into 60-mer in human BAFF is prevented by mutation of His218, a residue critical for 3-mer-to-3-mer interactions, but not for receptor binding. Despite the predominant functional role of processed BAFF in vivo, membrane-bound BAFF might also play a role. Indeed, soluble BAFF (3-mer) can trigger BAFF-R but not TACI or BCMA, whereas oligomeric forms of BAFF (BAFF 60-mer), which mimic membrane-bound BAFF, activate all BAFF receptors.

Fusion-Tag: His Tag

This recombinant proteins is for research use only.