Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant IL-2 Superkine (from HEK293 cells)
Human recombinant IL-2 Superkine (from HEK293 cells)
Catalog # PRSI90-293
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL-2 Superkine (from HEK293 cells)
Catalog # PRSI90-293
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein synonyms:
    Interleukin-2|IL-2 Superkine:Fc (rec.)|Super-2|Aldesleukin|TCGF|T Cell Growth Factor
  • Protein/peptide name:
    IL-2 Superkine
  • Purity:
    >95% (SDS-PAGE)
  • Sequence:
    21-153
  • Formulation:
    Lyophilized, Contains PBS.

Specifications

About this item

Interleukin-2 (IL-2) is a 133 amino acid glycoprotein with one intramolecular disulphide bond and variable glycosylation. It is secreted by activated T cells and induces proliferation and maturation of activated T cells, natural killer cells and lymphokine activated killer cells. IL-2 also stimulates proliferation of antibody-producing B cells, activates neutrophils and induces mononuclear cells to secrete IFN-gamma and TNF-alpha and -beta. Moreover, studies have shown that IL-2 is required for activation-induced apoptosis, an important homeostatic mechanism in the immune system, which is involved in the maintenance of peripheral tolerance to self-antigens. IL-2 promotes T cell proliferation and particularly naive T cells. IL-2 signalling on activated T cells is effected through a quaternary high-affinity receptor complex consisting of IL-2, IL-2Ralpha (CD25), IL-2Rbeta and IL-2Rgamma. Naive T cells are relatively insensitive to IL-2 as they only express small amounts of IL-2Rbeta and IL-2Rgamma. They only acquire sensitivity after CD25 expression, which captures the cytokine and presents it to the IL-2Rbeta and IL-2Rgamma receptors. IL-2 Superkine (Fc) is an artificial variant of IL-2 containing mutations at positions L80F / R81D / L85V / I 86V / I92F. These mutations are located in the molecule's core that acts to stabilize the structure and to give it a receptor-binding conformation mimicking native IL-2 bound to CD25. These mutations effectively eliminate the functional requirement of IL-2 for CD25 expression and elicit proliferation of T cells. Compared to IL-2, the IL-2 superkine induces superior expansion of cytotoxic T cells, leading to improved antitumor responses in vivo, and elicits proportionally less toxicity by lowering the expansion of Tregulatory cells and reducing pulmonary oedema.

Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.