Order Entry
Czech Republic
ContactUsLinkComponent
 
Mouse recombinant IL36 alpha (from E. coli)
  PRSI90-282
 :  
Mouse recombinant IL36 alpha (from E. coli)
  PRSI90-282
 :  

 

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Tag sequence:
    Tag Free
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at −20 °C.
  • Biological activity:
    Induces secretion of IL-6 in BMDCs at a concentration of 100ng - 1ug/ml.
  • Protein synonyms:
    Interleukin-1 Family Member 6|Interleukin-36alpha|IL-1H1|Interleukin-1epsilon|FIL1epsilon|IL-1F6|Interleukin-1 Homolog 1
  • Protein/peptide name:
    IL36 alpha
  • Purity:
    >95% (SDS-PAGE)
  • Sequence:
    Arg8-His160
  • Formulation:
    Lyophilized, Contains PBS.

 

 

IL-36alpha (IL-1F6), IL-36beta (IL-1F8) and IL-36gamma (IL-1F9) bind to IL-36R (IL-1Rrp2) and IL-1RAcP, activating similar intracellular signals as IL-1. IL-36Ra inhibits the production of proinflammatory cytokines, including IL-12, IL-1beta, IL-6, TNF-alpha and IL-23 induced by IL-36 in BMDC and CD4 T cells. Skin and dendritic cells are targets of the IL-36 interleukins leading to a Th1 response. These cytokines may represent potential targets for immune-mediated inflammatory conditions or, alternatively, could be used as adjuvants in vaccination.

Fusion-Tag: Tag Free

This recombinant proteins is for research use only.