Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant vaspin (from HEK293 cells)
Human recombinant vaspin (from HEK293 cells)
Catalog # PRSI90-059
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant vaspin (from HEK293 cells)
Catalog # PRSI90-059
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    DDDDK Tag
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at −20 °C.
  • Gene ID:
    145264
  • Protein synonyms:
    OL-64|Serpin A12|Visceral Adipose Tissue-derived Serine Protease Inhibitor
  • Protein/peptide name:
    Vaspin
  • Purity:
    >95% (SDS-PAGE)
  • Sequence:
    21-414
  • Formulation:
    Lyophilized, Contains PBS.

Specifications

About this item

Vaspin (Visceral adipose tissue-derived serpin; Serpin A12), a serine protease inhibitor (serpin), is an insulin-sensitising adipocytokine that has been isolated from both visceral and subcutaneous white adipose tissue. Based on recent findings, vaspin is suggested to regulate immune responses and inflammation and was found to be correlated with various metabolic parameters. Vaspin may represent a novel biomarker for obesity and impaired insulin sensitivity and might serve as a new therapeutic target of metabolic syndrome.

Fusion-Tag: DDDDK Tag

This recombinant proteins is for research use only.