Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Human recombinant FGF7 (from cells)

Human recombinant FGF7 (from cells)

50 µG
Catalog # PRSI92-679
Supplier:   ProSci Inc.
Human recombinant FGF7 (from cells)
Catalog # PRSI92-679
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant FGF7 (from cells)

50 µG

CAS Number:  
Price
18 530,00 Kč
Log in for availability
1 * 50 µG
Product Details & Documents

Fibroblast growth factor 7 (FGF7) is a secreted protein which is mainly located in epithelial cells and belongs to the heparin-binding growth factors family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumour growth and invasion. FGF7 is a potent epithelial cell-specific growth factor, whose mitogenic activity is predominantly exhibited in keratinocytes but not in fibroblasts and endothelial cells. It is possible major paracrine effector of normal epithelial cell proliferation.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Documents

Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    PRSI92-679
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    FGF7
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Protein synonyms:
    Keratinocyte growth factor|FGF7|Fibroblast growth factor 7|FGF-7|KGF|HBGF-7|Heparin-binding growth factor 7
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Tag sequence:
    C-6 His tag
  • Sequence:
    Cys32-Thr194
  • Species:
    Human
  • Source:
    Cells
  • Molecular weight:
    20 kD

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.