Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant LIGHT (from cells)
Human recombinant LIGHT (from cells)
Catalog # PRSI92-675
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant LIGHT (from cells)
Catalog # PRSI92-675
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    HVEM-L|Tumor necrosis factor ligand superfamily member 14|CD258|LIGHT|TNFSF14
  • Protein/peptide name:
    Light
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    18.2 kD
  • Sequence:
    Leu83-Val240
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Human TNFSF14 Protein, also known as LIGHT, belongs to a member of the tumour necrosis factor (TNF) ligand family. It can bind to NFRSF3/LTBR. It is a ligand for TNFRSF14, which is a member of the tumour necrosis factor receptor superfamily, and it is also known as a herpesvirus entry mediator ligand (HVEML). TNFSF14 encodes a protein with a 37 aa cytoplasmic domain, 21aa transmembrane domain and 182 aa extracellular region. The gene is predominantly expressed in the spleen and also found in the brain. Weakly expressed in peripheral lymphoid tissues and in heart, placenta, liver, lung, appendix, and kidney, and no expression seen in fetal tissues, endocrine glands, or nonhematopoietic tumour lines. TNFSF14 protein was found to probably function as a costimulatory factor for the activation of lymphoid cells and as a deterrent to infection by herpesvirus. Studies have shown that this protein can prevent tumour necrosis factor alpha mediated apoptosis in primary hepatocyte. Two alternatively spliced transcript variant encoding distinct isoforms have been reported.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.