Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant PDL1 (from cells)
Human recombinant PDL1 (from cells)
Catalog # PRSI92-671
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant PDL1 (from cells)
Catalog # PRSI92-671
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    PDL1|B7-H1|PD-L1|Programmed Cell Death 1 Ligand 1|B7H1|B7 Homolog 1|PDCD1 Ligand 1|CD274|PDCD1L1|Programmed Death Ligand 1|PDCD1LG1
  • Protein/peptide name:
    PDL1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    52.4 kD
  • Sequence:
    Phe19-Thr239
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

CD274, also known as B7-H1 or programmed death ligand 1 (PD-L1), is a 40 kD type I transmembrane protein and a member of the B7 family within the immunoglobulin receptor superfamily. Programmed death-1 ligand-1 (PD-L1, CD274, B7-H1) has been identified as the ligand for the immunoinhibitory receptor programmed death-1(PD1/PDCD1) and has been demonstrated to play a role in the regulation of immune responses and peripheral tolerance. By binding to PD1 on activated T-cells and B-cells, PD-L1 may inhibit ongoing T-cell responses by inducing apoptosis and arresting cell-cycle progression. Accordingly, it leads to growth of immunogenic tumour growth by increasing apoptosis of antigen specific T cells and may contribute to immune evasion by cancers. PD-L1 thus is regarded as promising therapeutic target for human autoimmune disease and malignant cancers.

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.