Order Entry
Czech Republic
ContactUsLinkComponent
Mouse recombinant IL-18 binding
Mouse recombinant IL-18 binding
Catalog # PRSI92-637
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant IL-18 binding
Catalog # PRSI92-637
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Interleukin 18 Binding Protein Isoform d|IL-18 BPd
  • Protein/peptide name:
    IL-18 binding
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    45.1 kD
  • Sequence:
    Thr29-Ala193
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Interleukin 18 binding protein (IL-18BP) is a physiological inhibitor that acts through binding to the receptor-binding site of IL-18. IL-18 stimulates INF- gamma , which then stimulates 18BP production via NF-κB. The interaction between IL-18 and IL-18BP has a significant role in the inflammation process. IL-18 BPs have four isoforms, a, b, c and d, which are spliced by different ways. The IL-18 BP isoforms a and c each contain one immunoglobulin (Ig)-like C2-type domain which is essential to the binding and neutralizing properties of the binding proteins. The IL-18 BP isoforms b and d lack a complete Ig domain. The expression of IL-18 and the IL-18BP are unidentified in immune tissues such as the spleen, but also in nonimmune cells including keratinocytes.

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.