Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant IL-15 R alpha & IL-15 fusion (from Cells)
Human recombinant IL-15 R alpha & IL-15 fusion (from Cells)
Catalog # PRSI92-588
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL-15 R alpha & IL-15 fusion (from Cells)
Catalog # PRSI92-588
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    IL-15RA|interleukin-15 receptor subunit alpha|IL-15R-alpha|IL-15 receptor subunit alpha|IL15|Interleukin-15|IL-15|IL15RA&IL15
  • Protein/peptide name:
    IL-15 R alpha & IL-15 fusion
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    46.9 kD
  • Sequence:
    Ile31-Asp96&Asn49-Ser162
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Interleukin-15 receptor subunit alpha, also known as Il15ra, is a high-affinity receptor for interleukin-15. Il15ra associates as a heterotrimer with the IL-2 receptor beta and gamma subunits (Common gamma chain, or gamma c) to initiate signal transduction. It can signal both in cis and trans where IL15R from one subset of cells presents IL15 to neighboring IL2RG-expressing cells. Il15ra is expressed in special cells including a wide variety of Tand B cells and non-lymphoid cells. Human Il15ra shares 45% amino acid sequence homology with the mouse form of the receptor. Eight isoforms of IL-15 R alpha mRNA have been identified, resulting from alternative splicing events involving different exons.Interleukin 15 (IL-15) is a cytokine that regulates T cell and natural killer cell activation and proliferation. IL-15 binds to the alpha subunit of the IL15 receptor (IL-15RA) with high affinity. IL-15 also binds to the beta and gamma chains of the IL-2 receptor, but not the alpha subunit of the IL2 receptor. IL-15 is structurally and functionally related to IL-2. Both cytokines share some subunits of receptors, allowing them to compete for and negatively regulate each other's activity. The number of CD8+ memory T cells is controlled by a balance between IL-15 and IL-2. Despite their many overlapping functional properties, IL-2 and IL-15 are, in fact, quite distinct players in the immune system. IL-15 is constitutively expressed by a wide variety of cell types and tissues, including monocytes, macrophages and DCs.

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.