Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Tag sequence:C-Fc tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:sIL-15 receptor subunit alpha|IL-15RA|Il15ra|Interleukin-15 receptor subunit alpha
- Protein/peptide name:IL-15 R alpha
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:45.5 kD
- Sequence:Gly33-Lys205
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Specifications
About this item
Mouse interleukin-15 receptor subunit alpha, also known as Il15ra, is a high-affinity receptor for interleukin-15. Il15ra associates as a heterotrimer with the IL-2 receptor beta and gamma subunits (Common gamma chain, or gamma c) to initiate signal transduction. It can signal both in cis and trans where IL15R from one subset of cells presents IL15 to neighboring IL2RG-expressing cells. Il15ra is expressed in special cells including a wide variety of Tand B cells and non-lymphoid cells. Human Il15ra shares 45% amino acid sequence homology with the mouse form of the receptor. Eight isoforms of IL-15 R alpha mRNA have been identified, resulting from alternative splicing events involving different exons.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.