Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant Cadherin-16 (from Cells)
Human recombinant Cadherin-16 (from Cells)
Catalog # PRSI92-418
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Cadherin-16 (from Cells)
Catalog # PRSI92-418
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    CDH16|CDH16/Cadherin-16/Kidney-specific cadherin/Ksp-cadherin/UNQ695/PRO1340
  • Protein/peptide name:
    Cadherin-16
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    84.6 kD
  • Sequence:
    Pro18-Ala786
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Cadherin-16(CDH16) is a single-pass type I membrane protein which contains six cadherin domains. Mature cadherin proteins consist of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small highly conserved C-terminal cytoplasmic domain. Cadherins are calcium-dependent cell adhesion proteins and may contribute to the sorting of heterogeneous cell types. They preferentially interact with themselves in a homophilic manner in connecting cells. Three calcium ions are usually bound at the interface of each cadherin domain and rigidify the connections, imparting a strong curvature to the full-length ectodomain. CDH16 is exclusively expressed in kidney, where the protein functions as the principal mediator of homotypic cellular recognition. It plays a role in the morphogenic direction of tissue development. CDH16 is composed of an extracellular domain containing 6 cadherin domains, a transmembrane region and a truncated cytoplasmic domain. However, it lacks the prosequence and tripeptide HAV adhesion recognition sequence typical of most classical cadherins.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.