Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant PRADC1 (from cells)
Human recombinant PRADC1 (from cells)
Katalogové číslo PRSI92-328
Dodavatel:  ProSci Inc.
Číslo CAS:  
Human recombinant PRADC1 (from cells)
Katalogové číslo PRSI92-328
Dodavatel:  ProSci Inc.
Číslo CAS:  

Specifikace

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    C2orf7|Protease-associated domain-containing protein of 21 kDa|PAP21|Protease-associated domain-containing protein 1|UNQ833/PRO1760|Hpap21
  • Protein/peptide name:
    PRADC1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    20 kD
  • Sequence:
    His22-Trp188
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifikace

O tomto produktu

PRADC1, also known as C2orf7 or PAP21, is short for Protease-associated domain-containing protein 1. It is a 188 aa. with a 21 aa. signal, and the 171 located Asn can be glycosylated. PRADC1 has two mutagenesis which are N121Q and N171Q. This protein is secreted and highly expressed in skeletal muscle, heart and liver. It is expressed at intermediate level in kidney.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.