Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant Ferritin light chain (from E. coli)
Human recombinant Ferritin light chain (from E. coli)
Catalog # PRSI92-265
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Ferritin light chain (from E. coli)
Catalog # PRSI92-265
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Ferritin L subunit|Ferritin light chain|FTL
  • Protein/peptide name:
    Ferritin light chain
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    24.5 kD
  • Sequence:
    Met1-Asp175
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Ferritin is a large, iron-storage heteropolymeric protein,which is expressed in most kinds of cells and co-assemble in different proportion in a tissue-specific manner. Ferritin has oligomer of 24 subunits and two types of subunits including light chain(FTL) and heavy chain. Ferritin can remove Fe (Ⅱ) from solution in the presence of oxygen and is very important for iron homeostasis. Iron is absorbed in the ferrous form and deposited as ferric hydroxides after oxidation. Iron is first oxidised to the ferric state for storage as ferric oxyhdroxide whithin the protein shell of ferritin. Thus, ferritin removes excess iron from the cell sap where it could otherwise participate in peroxidation mechanisms. Ferritin also plays a role in delivery of iron to cells and mediates iron uptake in capsule cells of the developing kidney.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.