Human recombinant parvulin-14 (from E. coli)
Catalog # PRSI92-173
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Peptidyl-prolyl cis/trans isomerase EPVH|PIN4|Parvulin-17|Peptidyl-prolyl cis-trans isomerase Pin4|Parvulin-14|Rotamase Pin4|Peptidyl-prolyl cis-trans isomerase NIMA-interacting 4
- Protein/peptide name:Parvulin-14
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:18.8 kD
- Sequence:Met1-Lys156
- Formulation:Supplied as a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Peptidyl-prolyl cis-trans isomerase NIMA-interacting 4(PIN4) is a peptidyl-prolyl cis/trans isomerase (PPIase) which interacts with NIMA and is vital for cell cycle regulation. PIN4 has 2 different isoforms: PAR14 and PAR17. Furthermore, PIN4 protein binds to double-stranded DNA under physiological salt conditions. PIN4 is involved as a ribosomal RNA processing factor in ribosome biogenesis. The PAR14 binds to tightly bent AT-rich stretches of double-stranded DNA, but PAR17 binds to double-stranded DNA.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.