Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant ETS1 (from E. coli)
Human recombinant ETS1 (from E. coli)
Catalog # PRSI92-104
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant ETS1 (from E. coli)
Catalog # PRSI92-104
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    EWSR2|V-ets Erythroblastosis Virus E26 Oncogene Homolog 1 (Avian)|ETS1|Protein C-ets-1|Isoform CRA_a|p54|hCG_1732038|ETS1 Protein
  • Protein/peptide name:
    ETS1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    33 kD
  • Sequence:
    Met1-Thr272
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl,1mM EDTA,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

ETS1 Protein (ETS1) is a nuclear protein that belongs to the ETS family. Members of this family recognize the core consensus DNA sequence GGAA/T in target genes. Proteins function either as transcriptional activators or repressors of numerous genes. They are involved in stem cell development, cell senescence and death, and tumorigenesis. ETS1 is a transcription factor, containing one ETS DNA-binding domain and one PNT (pointed) domain. it has been shown to interact with TTRAP, UBE2I and Death Associated Protein 6.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag