Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant PLA2G16 (from E. coli)
Human recombinant PLA2G16 (from E. coli)
Catalog # PRSI92-042
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant PLA2G16 (from E. coli)
Catalog # PRSI92-042
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    Group XVI Phospholipase A1/A2|HREV107-3|HRSL3|HREV107|HRASLS3|AdPLA|Renal Carcinoma Antigen NY-REN-65|H-Rev 107 Protein Homolog|PLA2G16|HRAS-like Suppressor 1|Adipose-Specific Phospholipase A2|HREV107-1|HRAS-like Suppressor 3
  • Protein/peptide name:
    PLA2G16
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    15.93 kD
  • Sequence:
    Asp12-Asp132
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    used for biological assays

Specifications

About this item

Group XVI Phospholipase A1/A2 (PLA2G16) belongs to the H-rev 107 family. PLA2G16 is expressed in a number of human tumors including ovarian carcinomas, lung carcinomas. PLA2G16 is involved in the regulation of differentiation and survival. PLA2G16 regulates adipocyte lipolysis and release of fatty acids through a G-protein coupled pathway involving prostaglandin and EP3. It has also been reported to play a crucial role in the development of obesity in mouse models.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag