Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant BCAS2 (from E. coli)
Human recombinant BCAS2 (from E. coli)
Catalog # PRSI92-031
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant BCAS2 (from E. coli)
Catalog # PRSI92-031
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His, N-T7 tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Pre-mRNA-Splicing Factor SPF27|DNA Amplified in Mammary Carcinoma 1 Protein|Spliceosome-Associated Protein SPF 27|BCAS2|DAM1|Breast Carcinoma-Amplified Sequence 2
  • Protein/peptide name:
    BCAS2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    28.58 kD
  • Sequence:
    Ala2-Phe225
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 200mM NaCl, 2mM DTT, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Breast Carcinoma-Amplified Sequence 2 (BCAS2) is a member of the SPF27 family. BCAS2 is a nuclear protein and widely expressed in many rtissues. BCAS2 is identified as being overexpressed in various breast cancer cell lines. BCAS2 is a component of the spliceosome, taking part in the removal of introns from mRNA precursors. BCAS2 interacts with estrogen receptor alpha and beta, thyroid hormone receptor beta, peroxisome proliferator-activated receptor gamma. BCAS2 functions as an ER co-activator and is capable of enhancing ER-mediated transcription.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His, N-T7 tag