Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Human recombinant Testin (from <i>E. coli</i>)

Human recombinant Testin (from E. coli)

50 µG
Catalog # PRSI92-024
Supplier:   ProSci Inc.
Human recombinant Testin (from <i>E. coli</i>)
Catalog # PRSI92-024
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Testin (from E. coli)

50 µG

CAS Number:  
Price
14 510,00 Kč
Log in for availability
1 * 50 µG
Product Details & Documents

Testin belongs to the prickle/espinas/testin family. Testin contains three LIM zinc-binding domains and one PET domain, TES as a scaffold protein that may have a role in cell adhesion, cell spreading and in the reorganization of the actin cytoskeleton. In addition, TES can also as a tumor suppressor, inhibits tumor cell growth, regulate the cell proliferation.TES interacts with many cytoskeletal proteins, such as Zyxin, Mena, Actin, Talin and VASP. The ability of TES to associate with alpha-actin, Zyxin and paxillin is dependent on the conformational state of the molecule.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

Documents

Specifications

Specifications

  • Pk:
    50 µG
  • Protein/peptide name:
    Testin
  • Protein synonyms:
    TES|TESS|Testin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Sequence:
    Met1-Ser421
  • Source:
    E. coli
  • Molecular weight:
    49.1 kD
  • Cat. No.:
    PRSI92-024
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 1mM DTT, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    C-6 His tag
  • Tested applications:
    used for biological assays
  • Species:
    Human

Specifications