Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant Autophagy 10 (from E. coli)
Human recombinant Autophagy 10 (from E. coli)
Catalog # PRSI91-998
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Autophagy 10 (from E. coli)
Catalog # PRSI91-998
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His, N-T7 tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    Ubiquitin-like-Conjugating Enzyme ATG10|Autophagy-Related Protein 10|APG10-like|ATG10
  • Protein/peptide name:
    Autophagy 10
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    24.5 kD
  • Sequence:
    Met1-Thr190
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Ubiquitin-Like-Conjugating Enzyme ATG10 (ATG10) is a ubiquitous 28kDa member of the ATG10 family protein. ATG10 acts as an E2-like enzyme, catalyzes the transfer of ATG12 to ATG5 during in the initial stages of autophagesome formation. The heterodimer of ATG5 and ATG12 subsequntly associates non-covalently with an ATG16 multimer to generate an antophagosome. ATG10 plays a role in the conjugation of ATG12 to ATG5 by interaction with MAP1LC3A. In addition, ATG10 can diretly interact with ATG5 or ATG7.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His, N-T7 tag