Human recombinant Peroxiredoxin 6 (from E. coli)
Catalog # PRSI91-974
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Red Blood Cells Page Spot 12|KIAA0106|PRDX6|Peroxiredoxin-6|24 kDa Protein|1-Cys PRX|AOP2|Liver 2D PAGE Spot 40|NSGPx|Acidic Calcium-Independent Phospholipase A2|Non-Selenium Glutathione Peroxidase|Antioxidant Protein 2|aiPLA2|1-Cys Peroxiredoxin
- Protein/peptide name:Peroxiredoxin 6
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:27.2 kD
- Sequence:Met1-Pro224
- Formulation:Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:used for biological assays
Specifications
About this item
Peroxiredoxin 6 belongs to the thiol-specific antioxidant protein family, which is a bifunctional enzyme with two distinct active sites. Peroxiredoxin 6 is involved in redox regulation of the cell and can reduce Hydrogen peroxide and short chain organic, fatty acid, and phospholipid hydroperoxides. Peroxiredoxin 6 may regulates phospholipid turnover and protectes against oxidative injury. Peroxiredoxin 6 eases the oxidative stress and TGF-beta-induced abnormalities of human trabecular meshwork cells, it is necessary for blood vessel integrity in injured skin.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag