Human recombinant Galectin 14 (from cells)
Catalog # PRSI91-644
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:CLC2|LGALS14|Charcot-Leyden Crystal Protein 2|PPL13|Gal-14|Placental Protein 13-like|Galectin-14
- Protein/peptide name:Galectin 14
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:17.1 kD
- Sequence:Met1-Asp139
- Formulation:Supplied as a 0.2 um filtered solution of 20mM Tri,100mM NaCl,1mM DTT,20% glycerol,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Galectin-14 is a member of the Galectin family of carbohydrate binding proteins. The members of Galectin family contain one or two carbohydrate recognition domains, which can bind beta -Galactoside. LGALS14 is expressed intracellularly in placenta and eosinophils, and is released by eosinophils following allergen stimulation. LGALS14 may be involved in the development of allergic inflammation. Two alternatively spliced transcript variants encoding distinct isoforms have been observed.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag