Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant S100 calcium binding A9 (from E. coli)
Human recombinant S100 calcium binding A9 (from E. coli)
Catalog # PRSI91-637
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant S100 calcium binding A9 (from E. coli)
Catalog # PRSI91-637
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Protein S100-A9|CAGB|MRP14|Calprotectin L1H subunit|CFAG|Leukocyte L1 complex heavy chain|Calgranulin-B|MRP-14
  • Protein/peptide name:
    S100 calcium binding A9
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    13.2 kD
  • Sequence:
    Thr2-Pro114
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Protein S100-A9 (also MRP14 and calgranulin B)is a calcium- and zinc-binding protein which plays a prominent role in the regulation of inflammatory processes and immune response.It can induce neutrophil chemotaxis, adhesion, can increase the bactericidal activity of neutrophils by promoting phagocytosis via activation of SYK, PI3K/AKT, and ERK1/2 and can induce degranulation of neutrophils by a MAPK-dependent mechanism.

  • This recombinant protein can be used for biological assays. For research use only.