Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant CXCL1 (from cells)
Human recombinant CXCL1 (from cells)
Catalog # PRSI91-532
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant CXCL1 (from cells)
Catalog # PRSI91-532
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    CXCL1|C-X-C Motif Chemokine 1|GRO|SCYB1|Growth-Regulated Alpha Protein|GRO1|Neutrophil-Activating Protein 3|Melanoma Growth Stimulatory Activity|GRO-Alpha(1-73)|NAP-3|MGSA|GRO alpha|GROA
  • Protein/peptide name:
    CXCL1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    8.9 kD
  • Sequence:
    Ala35-Asn107
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 5% Trehalose, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Chemokine (C-X-C motif) Ligand 1 Protein (CXCL1) is a growth factor for melanoma cells and a chemotaxin for neutrophils and a member of the CXC chemokine family that is a potent neutrophil attractant and activator and is also active toward basophils. CXCL1 is expressed by macrophages, neutrophils and epithelial cells; it has neutrophil chemoattractant activity. CXCL1 plays a critical nonredundant role in the development of experimental Lyme arthritis and carditis via CXCR2-mediated recruitment of neutrophils into the site of infection and may also have important pro-nociceptive effects via its direct actions on sensory neurons, and may induce long-term changes that involve protein synthesis.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag