Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant LOX-1 (from cells)
Human recombinant LOX-1 (from cells)
Catalog # PRSI91-474
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant LOX-1 (from cells)
Catalog # PRSI91-474
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Lectin-Type Oxidized LDL Receptor 1|LOX-1|hLOX-1|Lectin-like oxLDL Receptor 1|Ox-LDL Receptor 1|CLEC8A|Lectin-like Oxidized LDL Receptor 1|OLR1|LOX1|C-Type Lectin Domain Family 8 Member A|Oxidized Low-Density Lipoprotein Receptor 1
  • Protein/peptide name:
    LOX-1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    25.39 kD
  • Sequence:
    Ser61-Gln273
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Oxidized Low-Density Lipoprotein Receptor 1 (Ox-LDL Receptor 1) is a secreted, single-pass type II membrane protein which belongs to the C-type lectin superfamily. Ox-LDL Receptor 1 is expressed at high levels in endothelial cells and vascular-rich organs such as placenta, lung, liver, brain, aortic intima, bone marrow, spinal cord and substantia nigra. The expression of Ox-LDL Receptor 1 is induced by inflammatory cytokines such as TNF, IFNG and IL6 by pathological conditions, such as hyperlipidemia, hypertension and diabetes mellitus. Ox-LDL Receptor 1 mediates the recognition, internalization and degradation of oxidatively modified low density lipoprotein (OxLDL) by vascular endothelial cells. Ox-LDL Receptor 1 association with oxLDL induces the activation of NF-kappa-B through an increased production of intracellular reactive oxygen and a variety of pro-atherogenic cellular responses including a reduction of nitric oxide (NO) release, monocyte adhesion and apoptosis. Ox-LDL Receptor 1 also binds to oxLDL, which acts as a receptor for the HSP70 protein involved in antigen cross-presentation to naive T-cells in dendritic cells, thereby participating in cell-mediated antigen cross-presentation. It also participates in inflammatory process, by acting as a leukocyte-adhesion molecule at the vascular interface in endotoxin-induced inflammation.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag