Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant SP-10 (from cells)
Human recombinant SP-10 (from cells)
Catalog # PRSI91-378
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant SP-10 (from cells)
Catalog # PRSI91-378
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Acrosomal Vesicle Protein 1|Acrosomal Protein SP-10|ACRV1
  • Protein/peptide name:
    SP-10
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    26.88 kD
  • Sequence:
    Gln22-Ile265
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays

Specifications

About this item

Acrosomal Protein SP-10 is a testis-specific differentiation antigen that is associated with acrosomal membranes and matrix of mature sperm. It has been detected in several species including humans. Acrosomal Protein SP-10 may be involved in sperm-zona binding or penetration as a potential contraceptive vaccine immunogen for humans. ACRV1 is also a intra-acrosomal protein that is considered to be a vaccine candidate for immunocontraception.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag