Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Human recombinant CD86 (from cells)

Human recombinant CD86 (from cells)

50 µG
Catalog # PRSI91-371
Supplier:   ProSci Inc.
Human recombinant CD86 (from cells)
Catalog # PRSI91-371
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant CD86 (from cells)

50 µG

CAS Number:  
Price
9 240,00 Kč
Log in for availability
1 * 50 µG
Product Details & Documents

The protein is the receptor that involved in the costimulatory signal essential for T-lymphocyte proliferation and interleukin-2 production, by binding CD28 or CTLA-4. It may play a critical role in the early events of T-cell activation and costimulation of naive T-cells, such as deciding between immunity and anergy that is made by T-cells within 24 hours after activation. Isoform 2 interferes with the formation of CD86 clusters, and thus acts as a negative regulator of T-cell activation. The protein interacts with MARCH8, human herpesvirus 8 MIR2 protein, adenovirus subgroup B fiber proteins and acts as a receptor for these viruses.It is expressed by activated B-lymphocytes and monocytes and promoted by MARCH8 and results in endocytosis and lysosomal degradation.It contains 1 Ig-like C2-type(immunoglobulin-like) domainand 1 Ig-like V-type (immunoglobulin-like) domain.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

Documents

Specifications

Specifications

  • Pk:
    50 µG
  • Protein/peptide name:
    CD86
  • Protein synonyms:
    B70|T-Lymphocyte Activation Antigen CD86|BU63|CD86|Activation B7-2 Antigen|CTLA-4 Counter-Receptor B7.2|CD28LG2|FUN-1|B7-2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Sequence:
    Ala24-Pro247
  • Source:
    Cells
  • Molecular weight:
    26.69 kD
  • Cat. No.:
    PRSI91-371
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    C-6 His tag
  • Tested applications:
    For Biological Assays
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.