Human recombinant Kallikrein 6 (from cells)
Catalog # PRSI91-337
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:PRSS18|KLK6|Kallikrein-6|PRSS9|Neurosin|Zyme|Serine Protease 9|SP59|Protease M|Serine Protease 18
- Protein/peptide name:Kallikrein 6
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:26.17 kD
- Sequence:Glu17-Lys244
- Formulation:Supplied as a 0.2 um filtered solution of 5mM HCl, 150mM NaCl. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many Kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen Kallikrein subfamily members located in a cluster on chromosome 19. Its encoded protein is secreted and may play a role in suppression of tumorigenesis in breast and prostate cancers. Alternate splicing of this gene results in multiple transcript variants encoding the same protein.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag