Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant IFN alpha (from cells)
Human recombinant IFN alpha (from cells)
Catalog # PRSI91-329
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IFN alpha (from cells)
Catalog # PRSI91-329
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    IFN-Alpha/Beta Receptor 1|CRF2-1|beta R1|Type I Interferon Receptor 1|Cytokine Receptor Family 2 Member 1|IFNAR|Cytokine Receptor Class-II Member 1|IFNAR1|IFN-R-1|Interferon Alpha/Beta Receptor 1
  • Protein/peptide name:
    IFN alpha
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    48.18 kD
  • Sequence:
    Lys28-Lys436
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays

Specifications

About this item

The Interferon- alpha/ beta Receptor 1 (IFN- alpha/ beta R1) is a receptor which binds Type I Interferons including Interferon- alpha and - beta . It is a cell surface receptor and heteromeric receptor composed of one chain with two subunits referred to as IFNAR1 and IFNAR2. IFN- alpha/ beta R1, in association with IFN- alpha/ beta R2, is required for propagating antiviral signal transduction triggered by IFN- alpha and IFN- beta . IFN- alpha/ beta R1 interacts very weakly or not at all with type 1 interferons and does not stably interact with IFN- alpha/ beta R2. Ligands associate with IFN- alpha/ beta R2, and this complex subsequently forms a stable ternary assembly with IFN- alpha/ beta R1. IFN- alpha/ beta R1 also associates with IFN- gamma R2 even in the absence of IFN- gamma stimulation. Human IFN- alpha/ beta R1 contains a nuclear localization signal in its extracellular domain that is required for receptor translocation to the nucleus following interaction with ligand. Interferon stimulation results in an immunologic response that is especially associated with viruses.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag