Human recombinant IGF Binding 4 (from cells)
Catalog # PRSI91-320
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:IBP4|IGFBP4|IGFBP-4|IGF-Binding Protein 4|IBP-4|Insulin-like Growth Factor-Binding Protein 4
- Protein/peptide name:IGF Binding 4
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:27.01 kD
- Sequence:Asp22-Glu258
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
IGFBP-4, whose full name is insulin-like growth factor-binding protein 4, is induced by forskolin and N6, O2’dibutyryl sdenosine 3’, or 5’-cyclic monophosphate. It contains IGFBP N-terminal domain and thyroglobulin type-1 domain, and can bind IGF2. The IGF-binding proteins can prolong the half-life of the IGFs and have been shown to either inhibit or stimulate the growth promoting effects of the IGFs on cell culture.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag