- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:CRISP3|Cysteine-Rich Secretory Protein 3|Specific Granule Protein of 28 kDa|CRISP-3|SGP28
- Protein/peptide name:CRISP3
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:26.54 kD
- Sequence:Asn21-Tyr245
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:For Biological Assays
Cysteine-rich secretory protein 3 (CRISP-3) is a secreted protein,containing 1 SCP domain and 1 ShKT domain.It is belongs to the CRISP family. CRISP-3 is a glycoprotein that belongs to the family of cysteine-rich secretory proteins (CRISPs) which was originally discovered in human neutrophilic granulocytes. CRISP-3 is also widely distibuted in exocrine glands (salivary glands, pancreas and prostate), eosinophilic granulocytes and to a lower level in epididymis, ovary, thymus and colon. The presence of CRISP-3 in neutrophils, eosinophils and in exocrine secretions indicates a role in innate host defense. The antibody has been raised against recombinant C-terminally truncated form of CRISP-3 and recognizes both the N-glycosylated and non-glycosylated form of the mature protein.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag