Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant LDHA (from E. coli)
Human recombinant LDHA (from E. coli)
Catalog # PRSI91-215
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant LDHA (from E. coli)
Catalog # PRSI91-215
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    Cell Proliferation-Inducing Gene 19 Protein|LDH-M|LDHA|L-Lactate Dehydrogenase A Chain|PIG19|LDH-A|Renal Carcinoma Antigen NY-REN-59|LDH Muscle Subunit
  • Protein/peptide name:
    LDHA
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    38.8 kD
  • Sequence:
    Met1-Phe332
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 50% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological assays

Specifications

About this item

L-Lactate Dehydrogenase A Chain (LDHA) is an enzyme that catalyzes the conversion of L-lactate and NAD+ to pyruvate and NADH in the final step of anaerobic glycolysis. LDHA contains an N-terminal coenzyme binding region, a central catalytic site, and at least nine utilized Lys acetylation and two Tyr phosphorylation sites. LDHA belongs to the lactate dehydrogenase family, expressed predominantly in muscle tissue. LDHA mutations have been linked to exertional myoglobinuria.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag