Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant PDGF-BB (from E. coli)
Human recombinant PDGF-BB (from E. coli)
  PRSI91-179
 :  
Human recombinant PDGF-BB (from E. coli)
  PRSI91-179
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    SIS|PDGF2|Platelet-Derived Growth Factor B Chain|Platelet-Derived Growth Factor Subunit B|Proto-Oncogene c-Sis|PDGF Subunit B|PDGFB|Platelet-Derived Growth Factor Beta Polypeptide|Becaplermin|PDGF-2
  • Protein/peptide name:
    PDGF-BB
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    12.42 kD
  • Sequence:
    Ser82-Thr190
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 4mM HCl. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Platelet-Derived Growth Factor Subunit B (PDGFB) belongs to the PDGF/VEGF growth factor family. Platelet-derived growth factor is a potent mitogen for cells of mesenchymal origin. PDGFB can exist either as a homodimer (PDGF-BB) or as a heterodimer with the platelet-derived growth factor alpha polypeptide (PDGF-AB), where the dimers are connected by disulfide bonds. Mutations in this gene are associated with meningioma.Binding of PDGFB to its receptor elicits a variety of cellular responses. In addition, PDGFB is released by platelets upon wounding and plays an important role in stimulating adjacent cells to grow and thereby heals the wound.

  • This recombinant protein can be used for biological assays. For research use only.