Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant Thioredoxin 2 (from E. coli)
Human recombinant Thioredoxin 2 (from E. coli)
Catalog # PRSI91-162
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Thioredoxin 2 (from E. coli)
Catalog # PRSI91-162
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Thioredoxin-2|TRX2|Thioredoxin Mitochondrial|Mt-Trx|MTRX|TXN2
  • Protein/peptide name:
    Thioredoxin 2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    12 kD
  • Sequence:
    Thr60-Gly166
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological assays

Specifications

About this item

Thioredoxin-2 (TXN2) is a mitochondrial member of the thioredoxin family. Thioredoxin-2 is extensively expressed in adult and fetal tissues. Thioredoxin-2 contains an N-terminal 59 amino acid transit peptide, which is cleaved before translocating to mitochondria. Mitochondrial thioredoxin play important roles in the regulation of the mitochondrial membrane potential and in protection against oxidant-induced apoptosis. Thioredoxin-2 could be involved in the resistance to anti-tumor agents and possesses a dithiol-reducing activity. In addition, Thioredoxin-2 is important at low oxidative stress conditions.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag