Order Entry
Czech Republic
ContactUsLinkComponent
Human Recombinant 40S Ribosomal S19 (from E. coli)
Human Recombinant 40S Ribosomal S19 (from E. coli)
Catalog # PRSI91-150
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant 40S Ribosomal S19 (from E. coli)
Catalog # PRSI91-150
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    RPS19|40S Ribosomal Protein S19
  • Protein/peptide name:
    40S Ribosomal S19
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    16.1 kD
  • Sequence:
    Pro2-His145
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

40S Ribosomal Protein S19 (RPS19) is a ribosomal protein that Belongs to the ribosomal protein S19e family. RPS19 is located in the nucleoli, and higher level expression is seen in colon carcinoma tissue than normal colon tissue. It required for pre-rRNA processing and maturation of 40S ribosomal subunits. RPS19 plays a role in many biological processes, such as endocrine pancreas development, erythrocyte differentiation, mRNA metabolic process. Defects in RPS19 are the cause of Diamond-Blackfan anemia type 1 (DBA1), which is a form of Diamond-Blackfan anemia, a congenital non-regenerative hypoplastic anemia that usually presents early in infancy. Diamond-Blackfan anemia is characterized by a moderate to severe macrocytic anemia, erythroblastopenia, and an increased risk of malignancy.

  • This recombinant protein can be used for biological assays. For research use only.